Invitrogen
Manufacturer Code:PA5104114
Catalog # PIPA5104114
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BMS1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA408648 ACC AU020092 AU043373 BB007109 BC030906 BMS1 BMS1 homolog ribosome assembly protein BMS1 homolog ribosome assembly protein (yeast) BMS1 ribosome biogenesis factor BMS1 ribosome biogenesis factor BMS1L BMS1-like ribosome assembly protein KIAA0187 mKIAA0187 Ribosome assembly protein BMS1 homolog ribosome biogenesis protein BMS1 homolog; BMS1 homolog, ribosome assembly protein; BMS1 ribosome biogenesis factor; BMS1-like, ribosome assembly protein; Ribosome assembly protein BMS1 homolog; Ribosome biogenesis protein BMS1 homolog
Gene Aliases: ACC; BMS1; BMS1L; KIAA0187
UniProt ID: (Human) Q14692
Entrez Gene ID: (Human) 9790
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.