Abnova
Manufacturer Code:H00000659M02
Catalog # 89-016-350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C10 |
Immunogen | BMPR2 (NP_001195.2 939 a.a. ~ 1037 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant BMPR2.
For Research Use Only
Protein Aliases: BMP type II receptor; BMP type-2 receptor; BMPR-2; BMPR-II; BMPR3; BMR2; Bone morphogenetic protein receptor type II; Bone morphogenetic protein receptor type-2; bone morphogenetic protein receptor, type II (serine/threonine kinase); BRK-3; FLJ41585; FLJ76945; PPH1; T-ALK; type II activin receptor-like kinase; type II receptor for bone morphogenetic protein-4
Gene Aliases: BMPR-II; BMPR2; BMPR3; BMR2; BRK-3; POVD1; PPH1; T-ALK
UniProt ID: (Human) Q13873
Entrez Gene ID: (Human) 659
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.