Invitrogen
Manufacturer Code:PA599465
Catalog # PIPA599465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BMPR2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610024H22Rik AL117858 AW546137 BB189135 BMP type II receptor BMP type-2 receptor BMP-2 Bmpr2 BMPR-2 BMPR3 BMPRII BMPR-II BMR2 bone morphogenetic protein receptor type 2 bone morphogenetic protein receptor type II Bone morphogenetic protein receptor type-2 bone morphogenetic protein receptor type II (serine/threonine kinase) bone morphogenic protein receptor type II (serine/threonine kinase) BRK-3 Gm20272 POVD1 PPH1 T-ALK type II activin receptor-like kinase type II receptor for bone morphogenetic protein-4; BMP type II receptor; BMP type-2 receptor; BMPR-2; Bone morphogenetic protein receptor type II; Bone morphogenetic protein receptor type-2; bone morphogenetic protein receptor, type II (serine/threonine kinase); bone morphogenic protein receptor, type II (serine/threonine kinase); BRK-3; type II activin receptor-like kinase; type II receptor for bone morphogenetic protein-4
Gene Aliases: 2610024H22Rik; AL117858; AW546137; BB189135; BMP-2; BMPR-2; BMPR-II; BMPR2; BMPR3; BMPRII; BMR2; BRK-3; Gm20272; POVD1; PPH1; T-ALK
UniProt ID: (Human) Q13873, (Mouse) O35607
Entrez Gene ID: (Human) 659, (Rat) 140590, (Mouse) 12168
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.