Invitrogen
Manufacturer Code:PA511863
Catalog # PIPA511863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 28-59 amino acids from the N-terminal region of human BMPR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP type II receptor; BMP type-2 receptor; BMPR-2; Bone morphogenetic protein receptor type II; Bone morphogenetic protein receptor type-2; Bone morphogenetic protein receptor type-2 Bone morphogenetic protein receptor type II BMP type II receptor BMPR-II PPH1; bone morphogenetic protein receptor, type II (serine/threonine kinase); bone morphogenic protein receptor, type II (serine/threonine kinase); BRK-3; type II activin receptor-like kinase; type II receptor for bone morphogenetic protein-4
Gene Aliases: 2610024H22Rik; AL117858; AW546137; BB189135; BMP-2; BMPR-2; BMPR-II; BMPR2; BMPR3; BMPRII; BMR2; BRK-3; Gm20272; POVD1; PPH1; T-ALK
UniProt ID: (Human) Q13873, (Mouse) O35607
Entrez Gene ID: (Human) 659, (Mouse) 12168
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.