Invitrogen
Manufacturer Code:MA517036
Catalog # PIMA517036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4B7B2 |
Immunogen | Purified recombit fragment of human BMPR1A (amino acids: 179-378 ) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor, type II-like kinase 3; Activin receptor-like kinase 3; ALK-3; BMP type-1A receptor; BMPR-1A; BMPRIA 10q23del ACVRLK3 ALK3 CD292 SKR5 ALK-3 BMP type-1A receptor BMPR-1A activin A receptor type II-like kinase 3 activin receptor-like kinase 3 bone morphogenetic protein receptor type-1A serine/threonine-protein kinase receptor R5; bone morphogenetic protein receptor type IA; Bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5
Gene Aliases: 10q23del; ACVRLK3; ALK3; BMPR1A; CD292; SKR5
UniProt ID: (Human) P36894
Entrez Gene ID: (Human) 657
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.