Abnova
Manufacturer Code:H00000657A03
Catalog # 89-104-162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 532 a.a) full-length recombinant protein with GST tag.,BMPR1A (AAH28383 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant BMPR1A.
For Research Use Only
Protein Aliases: 10q23del; activin A receptor, type II-like kinase 3; Activin receptor-like kinase 3; ACVRLK3; ALK-3; ALK3; BMP type-1A receptor; BMPR-1A; bone morphogenetic protein receptor type IA; Bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5
Gene Aliases: 10q23del; ACVRLK3; ALK3; BMPR1A; CD292; SKR5
UniProt ID: (Human) P36894
Entrez Gene ID: (Human) 657
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.