R&D Systems
Manufacturer Code:AF811
Catalog # AF811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human BMPR-II Ala26-Ile151 Accession # Q13873 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP type II receptor; BMP type II receptor BMP type-2 receptor BMPR2 BMPR-2 BMPR3 BMPRII BMPR-IIprimary pulmonary hypertension 1 Bone morphogenetic protein receptor type II bone morphogenetic protein receptor type-2 bone morphogenetic protein receptor type II (serine/threonine kinase) BRK-3 EC 2.7.11 EC 2.7.11.30 FLJ41585 FLJ76945 PPH1BMR2 T-ALK type II activin receptor-like kinase type II receptor for bone morphogenetic protein-4; BMP type-2 receptor; BMPR-2; Bone morphogenetic protein receptor type II; Bone morphogenetic protein receptor type-2; bone morphogenetic protein receptor, type II (serine/threonine kinase); type II activin receptor-like kinase; type II receptor for bone morphogenetic protein-4
Gene Aliases: BMPR-II; BMPR2; BMPR3; BMR2; BRK-3; POVD1; PPH1; T-ALK
UniProt ID: (Human) Q13873
Entrez Gene ID: (Human) 659
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.