Novus Biologicals
Manufacturer Code:NBP290398B
Catalog # NB073063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse BMPR-IA/ALK-3 (NP_033888.2 Met 1-Arg 152). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor, type II-like kinase 3; Activin receptor-like kinase 3; ACVRLK310q23del ALK-3 ALK3EC 2.7.11.30 BMP type-1A receptor BMPR-1A bone morphogenetic protein receptor type-1A bone morphogenetic protein receptor type IA CD292 CD292 antigen EC 2.7.11 Serine/threonine-protein kinase receptor R5 SKR5 type II-like kinase 3; ALK-3; BMP type-1A receptor; BMPR-1A; bone morphogenetic protein receptor type IA; Bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5
Gene Aliases: 10q23del; ACVRLK3; ALK3; BMPR1A; CD292; SKR5
UniProt ID: (Human) P36894
Entrez Gene ID: (Human) 657
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.