Novus Biologicals
Manufacturer Code:NBP215601
Catalog # NBP215601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human BMPR1A. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor, type II-like kinase 3; Activin receptor-like kinase 3; ACVRLK310q23del ALK-3 ALK3EC 2.7.11.30 BMP type-1A receptor BMPR-1A bone morphogenetic protein receptor type-1A bone morphogenetic protein receptor type IA CD292 CD292 antigen EC 2.7.11 Serine/threonine-protein kinase receptor R5 SKR5 type II-like kinase 3; ALK-3; BMP type-1A receptor; BMPR-1A; bone morphogenetic protein receptor type IA; Bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5
Gene Aliases: 10q23del; ACVRLK3; ALK3; BMPR1A; CD292; SKR5
UniProt ID: (Human) P36894
Entrez Gene ID: (Human) 657
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.