Invitrogen
Manufacturer Code:MA535530
Catalog # PIMA535530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1025 |
Immunogen | A synthesized peptide derived from human BMP6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: BMP BMP 6 BMP6 Bmp-6 Bone morphogenetic protein Bone morphogenetic protein 6 D13Wsu115e VG-1-R VG-1-related protein VGR Vgr1 VGR-1; BMP-6; Bone morphogenetic protein 6; vegetal related growth factor (TGFB-related); vegetal-related (TGFB related) cytokine; VG-1-R; VG-1-related protein; Vg1-related sequence; VGR-1
Gene Aliases: Bmp-6; BMP6; D13Wsu115e; VGR; VGR1
UniProt ID: (Human) P22004, (Mouse) P20722, (Rat) Q04906
Entrez Gene ID: (Human) 654, (Mouse) 12161, (Rat) 25644
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.