Proteintech
Manufacturer Code:132531AP150UL
Catalog # 50-172-8604
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BMP5 Fusion Protein Ag3952 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP 5 BMP5 bone morphogenetic protein 5; BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein 5; short ear
Gene Aliases: AU023399; Bmp-5; BMP5; se
UniProt ID: (Human) P22003, (Mouse) P49003
Entrez Gene ID: (Human) 653, (Mouse) 12160
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.