Abnova
Manufacturer Code:PAB9481
Catalog # 89-134-170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human BMP4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant BMP4.
For Research Use Only
Protein Aliases: BMP-2B; BMP-4; BMP2B BMP2B1 MCOPS6 ZYME; Bone morphogenetic protein 2B; Bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein
Gene Aliases: BMP2B; BMP2B1; BMP4; DVR4; MCOPS6; OFC11; ZYME
UniProt ID: (Human) P12644
Entrez Gene ID: (Human) 652
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.