Proteintech
Manufacturer Code:124921AP150UL
Catalog # 50-172-8020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BMP4 Fusion Protein Ag3191 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP 2B BMP 4 BMP2B BMP2B1 BMP4 Bone morphogenetic protein 2B bone morphogenetic protein 4 DVR4 MCOPS6 ZYME; BMP-2B; BMP-4; Bone morphogenetic protein 2B; Bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein
Gene Aliases: Bmp-4; BMP2B; Bmp2b-1; BMP2B1; BMP4; BOMPR4A; Dvr-4; DVR4; MCOPS6; OFC11; ZYME
UniProt ID: (Human) P12644, (Mouse) P21275
Entrez Gene ID: (Human) 652, (Mouse) 12159, (Rat) 25296
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.