Proteintech
Manufacturer Code:189821AP150UL
Catalog # 50-173-2303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP 15 BMP15 bone morphogenetic protein 15 GDF 9B GDF9B ODG2 POF4; BMP-15; Bone morphogenetic protein 15; GDF-9B; Growth/differentiation factor 9B
Gene Aliases: AU015375; AU018861; AU021453; Bmp-15; BMP15; C86824; C87336; GDF-9B; GDF9B; ODG2; POF4
UniProt ID: (Human) O95972, (Mouse) Q9Z0L4
Entrez Gene ID: (Human) 9210, (Mouse) 12155
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.