Invitrogen
Manufacturer Code:PA577881
Catalog # PIPA577881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human BMP8b. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-8; BMP-8B; bone morphogenetic protein 8 (osteogenic protein 2); Bone morphogenetic protein 8B; dJ118J21.1 (bone morphogenetic protein 8 (osteogenic protein 2)); OP-2; OP2 bone morphogenetic protein 8b BMP8B BMP8; Osteogenic protein 2
Gene Aliases: BMP8; BMP8B; OP2; Op3
UniProt ID: (Human) P34820, (Mouse) P55105
Entrez Gene ID: (Human) 656, (Mouse) 12164
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.