Novus Biologicals
Manufacturer Code:NBP104259AF750
Catalog # NP10429A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 40000000 |
Immunogen | Recombinant human BMP7 (293-431 aa) purified from E. coli |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: BMP-7; BMP-7 bone morphogenetic protein 7 Eptotermin alfa OP-1OP1 Osteogenic protein 1; Bone morphogenetic protein 7; Eptotermin alfa; OP-1; Osteogenic protein 1
Gene Aliases: BMP7; OP-1; OP1
UniProt ID: (Human) P18075
Entrez Gene ID: (Human) 655
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.