Invitrogen
Manufacturer Code:PA540524
Catalog # PIPA540524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human BMP7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-7; BMP7; bone moorphogenic protein-7; Bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; Eptotermin alfa; OP-1; Osteogenic protein 1
Gene Aliases: BMP-7; BMP7; OP-1; OP1
UniProt ID: (Human) P18075, (Mouse) P23359
Entrez Gene ID: (Horse) 100050299, (Rabbit) 100337873, (Human) 655, (Sheep) 443175, (Bovine) 540595, (Dog) 477270, (Guinea pig) 100725543, (Mouse) 12162, (Rat) 85272
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.