Novus Biologicals
Manufacturer Code:NB10062399
Catalog # NB10062399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the amino acids 1 - 14 of the native molecule conjugated to Keyhole limpet Haemocyanin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-7; BMP-7 bone morphogenetic protein 7 Eptotermin alfa OP-1OP1 Osteogenic protein 1; Bone morphogenetic protein 7; Eptotermin alfa; OP-1; Osteogenic protein 1
Gene Aliases: BMP7; OP-1; OP1
UniProt ID: (Human) P18075
Entrez Gene ID: (Human) 655
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.