Invitrogen
Manufacturer Code:PA547829
Catalog # PIPA547829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit mouse BMP-6 Ser372-His510 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-6; Bone morphogenetic protein 6; VG-1-related protein; VGR VGR1 BMP-6 VG-1-R VG-1-related protein VGR-1 Vg1-related sequence vegetal related growth factor (TGFB-related) vegetal-related (TGFB related) cytokine; VGR-1
Gene Aliases: Bmp-6; Bmp6; D13Wsu115e; Vgr1
UniProt ID: (Mouse) P20722
Entrez Gene ID: (Mouse) 12161
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.