Invitrogen
Manufacturer Code:PA511719
Catalog # PIPA511719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 25-55 amino acids from the N-terminal region of human Bmp6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-6; Bone morphogenetic protein 6; vegetal related growth factor (TGFB-related); vegetal-related (TGFB related) cytokine; VG-1-R; VG-1-related protein; Vg1-related sequence; VGR-1
Gene Aliases: BMP6; VGR; VGR1
UniProt ID: (Human) P22004
Entrez Gene ID: (Human) 654
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.