Invitrogen
Manufacturer Code:PA570633
Catalog # PIPA570633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of rat BMP-5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein 5; MGC34244; short ear
Gene Aliases: AU023399; Bmp-5; BMP5; bmp5l; hm:zehl0669; se; zehl0669; zgc:64230
UniProt ID: (Human) P22003, (Mouse) P49003
Entrez Gene ID: (Rabbit) 100344917, (Human) 653, (Dog) 474944, (Horse) 100056806, (Bovine) 507682, (Mouse) 12160, (Guinea pig) 100729769, (Rat) 315824, (Zebrafish) 394026
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.