R&D Systems
Manufacturer Code:MAB1876SP
Catalog # MAB1876SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 203706 |
Immunogen | <EM>E. coli</EM>-derived recombinant human BMP-3 Gln363-Arg472 Accession # P12645.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BMP-3; BMP-3A; BMP3 BMP-3 BMP3A BMP-3A bone morphogenetic protein 3 bone morphogenetic protein 3 (osteogenic) Bone morphogenetic protein 3A bone morphogenetic protein-3 osteogenin; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); Bone morphogenetic protein 3A; bone morphogenetic protein-3; Osteogenin
Gene Aliases: BMP-3A; BMP3; BMP3A
UniProt ID: (Human) P12645
Entrez Gene ID: (Human) 651
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.