Novus Biologicals
Manufacturer Code:NBP120894
Catalog # NBP120894
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-GHRNFPTGTV PKWDSH corresponding to internal region according to NP_001192.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-3; BMP-3 BMP3A BMP-3A bone morphogenetic protein 3 bone morphogenetic protein 3 (osteogenic) Bone morphogenetic protein 3A bone morphogenetic protein-3 osteogenin; BMP-3A; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); Bone morphogenetic protein 3A; bone morphogenetic protein-3; Osteogenin
Gene Aliases: BMP-3A; BMP3; BMP3A
UniProt ID: (Human) P12645
Entrez Gene ID: (Human) 651
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.