Novus Biologicals
Manufacturer Code:NBP289490G
Catalog # NB107866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 110 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human BMP-2 (NP_001191.1 Gln283-Arg396). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: BMP-2; BMP-2 BMP-2A BMP2ABone morphogenetic protein 2A bone morphogenetic protein 2; BMP-2A; Bone morphogenetic protein 2; Bone morphogenetic protein 2A
Gene Aliases: BDA2; BMP2; BMP2A
UniProt ID: (Human) P12643
Entrez Gene ID: (Human) 650
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.