Invitrogen
Manufacturer Code:MA523763
Catalog # PIMA523763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 100221 |
Immunogen | CHO-derived recombit human BMP-2 Gln283-Arg396 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2610024H22Rik AL117858 AW546137 BB189135 BMPR-II BMPRII; BMP-2; BMP-2A; Bone morphogenetic protein 2; Bone morphogenetic protein 2A
Gene Aliases: BDA2; BMP2; BMP2A
UniProt ID: (Human) P12643
Entrez Gene ID: (Human) 650
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.