Invitrogen
Manufacturer Code:PA578874
Catalog # PIPA578874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human BMP-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-2; BMP-2A; Bone morphogenetic protein 2; Bone morphogenetic protein 2 BMP-2 Bone morphogenetic protein 2A BMP-2A BMP2 BMP2A; Bone morphogenetic protein 2A
Gene Aliases: BDA2; Bmp-2; BMP2; BMP2A
UniProt ID: (Human) P12643, (Rat) P49001
Entrez Gene ID: (Human) 650, (Rat) 29373
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.