Novus Biologicals
Manufacturer Code:NBP261935AF488
Catalog # NB261935488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 28A |
Immunogen | Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human GDF9B (BMP15) |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: BMP-15; BMP-15 bone morphogenetic protein 15 GDF-9B GDF9BODG2 Growth/differentiation factor 9B POF4; Bone morphogenetic protein 15; GDF-9B; Growth/differentiation factor 9B
Gene Aliases: BMP15; GDF9B; ODG2; POF4
UniProt ID: (Human) O95972
Entrez Gene ID: (Human) 9210
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.