R&D Systems
Manufacturer Code:AF5917
Catalog # AF5917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse BMP-15/GDF-9B Gln268-Arg392 Accession # Q9Z0L4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: BMP-15; BMP15 BMP-15 bone morphogenetic protein 15 GDF-9B GDF9BODG2 Growth/differentiation factor 9B POF4; Bone morphogenetic protein 15; GDF-9B; Growth/differentiation factor 9B
Gene Aliases: BMP15; GDF9B; ODG2; POF4
UniProt ID: (Human) O95972
Entrez Gene ID: (Human) 9210
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.