Proteintech
Manufacturer Code:257221AP150UL
Catalog # 50-173-5037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BLOC1S3 Fusion Protein Ag22801 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 3; Biogenesis of lysosome-related organelles complex 1 subunit 3; biogenesis of lysosome-related organelles complex-1, subunit 3; BLOC 1 subunit 3 BLOC1S3 BLOS3 HPS8 RP; BLOC-1 subunit 3; reduced pigmentation, mouse, homolog of
Gene Aliases: BLOC1S3; BLOS3; HPS8; RP
UniProt ID: (Human) Q6QNY0
Entrez Gene ID: (Human) 388552
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.