Invitrogen
Manufacturer Code:PA525452
Catalog # PIPA525452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 14-40 amino acids from the central region of human BLOC1S2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11 kDa centrosome associated protein; biogenesis of lysosomal organelles complex-1, subunit 2; Biogenesis of lysosome-related organelles complex 1 subunit 2; biogenesis of lysosome-related organelles complex-1, subunit 2; biogenesis of lysosome-related organelles complex-1, subunit 2A; BLOC-1 subunit 2; BLOS2 RP11-316M21.4 BLOC-1 subunit 2 biogenesis of lysosome-related organelles complex 1 subunit 2 centrosomal 10 kDa protein centrosome protein oncogene centrosome-associated protein; centrosomal 10 kDa protein; centrosome protein oncogene; Centrosome-associated protein
Gene Aliases: 2410089B13Rik; BLOC1S2; Bloc1s2a; BLOS2; BORCS2; CEAP; CEAP11
UniProt ID: (Human) Q6QNY1, (Mouse) Q9CWG9
Entrez Gene ID: (Human) 282991, (Mouse) 73689
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.