Novus Biologicals
Manufacturer Code:NBP300218UV
Catalog # NB091717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant BLOC1S2 (Q6QNY1-2 Met1-Arg99). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11 kDa centrosome associated protein; biogenesis of lysosomal organelles complex-1 subunit 2 BLOC-1 subunit 2 CEAP centrosomal 10 kDa protein centrosome protein oncogene Centrosome-associated protein subunit 2; biogenesis of lysosomal organelles complex-1, subunit 2; Biogenesis of lysosome-related organelles complex 1 subunit 2; BLOC-1 subunit 2; centrosomal 10 kDa protein; centrosome protein oncogene; Centrosome-associated protein
Gene Aliases: BLOC1S2; BLOS2; BORCS2; CEAP; CEAP11
UniProt ID: (Human) Q6QNY1
Entrez Gene ID: (Human) 282991
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.