Abnova
Manufacturer Code:H00282991K
Catalog # 89-207-320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human BLOC1S2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human BLOC1S2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 11 kDa centrosome associated protein; biogenesis of lysosomal organelles complex-1, subunit 2; Biogenesis of lysosome-related organelles complex 1 subunit 2; BLOC-1 subunit 2; BLOS2 FLJ30135 MGC10120 RP11-316M21.4; centrosomal 10 kDa protein; centrosome protein oncogene; Centrosome-associated protein
Gene Aliases: BLOC1S2; BLOS2; BORCS2; CEAP; CEAP11
UniProt ID: (Human) Q6QNY1
Entrez Gene ID: (Human) 282991
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.