Proteintech
Manufacturer Code:196871AP150UL
Catalog # 50-173-2487
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 1; Biogenesis of lysosome-related organelles complex 1 subunit 1; biogenesis of lysosome-related organelles complex-1, subunit 1; BLOC 1 subunit 1 BLOC1S1 BLOS1 GCN5 like protein 1 GCN5L1 MICoA Protein RT14 RT14; BLOC subunit 1; BLOC-1 subunit 1; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 1; GCN5 general control of amino acid synthesis-like 1; GCN5 general control of amino-acid synthesis 5-like 1; GCN5-like protein 1; general control of amino acid synthesis, yeast homolog-like 1; general control of amino acid synthesis-like 1; MTA1-interacting coactivator; Protein RT14
Gene Aliases: AI839753; BLOC1S1; BLOS1; BORCS1; GCN5L1; MICoA; RT14
UniProt ID: (Human) P78537, (Mouse) O55102, (Rat) D3ZKU7
Entrez Gene ID: (Human) 2647, (Mouse) 14533, (Rat) 288785
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.