R&D Systems
Manufacturer Code:MAB36081
Catalog # MAB36081
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 646702 |
Immunogen | <EM>E. coli</EM>-derived recombinant human BLIMP1/PRDM1 Lys667-Cys789 Accession # O75626 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B-lymphocyte-induced maturation protein 1; Beta-interferon gene positive regulatory domain I-binding factor; Beta-interferon gene positive regulatory domain I-binding factor beta-interferon gene positive-regulatory domain I binding factor BLIMP-1 BLIMP1MGC118925 blmp-1 B-lymphocyte-induced maturation protein 1 MGC118922 MGC118923 Positive regulatory domain I-binding factor 1 PR domain containing 1 with ZNF domain PR domain zinc finger protein 1 PR domain-containing protein 1 PRDI-BF1MGC118924 PRDI-binding factor 1 PRDI-binding factor-1 PRDM1 PR-domain zinc finger protein 1 ZNFPR1A1; beta-interferon gene positive-regulatory domain I binding factor; BLIMP-1; Positive regulatory domain I-binding factor 1; PR domain containing 1, with ZNF domain; PR domain zinc finger protein 1; PR domain-containing protein 1; PRDI-BF1; PRDI-binding factor-1
Gene Aliases: BLIMP1; PRDI-BF1; PRDM1
UniProt ID: (Human) O75626
Entrez Gene ID: (Human) 639
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.