OriGene
Manufacturer Code:TA500761
Catalog # 50-167-1903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B10 |
Immunogen | Full length human recombit protein of human BIRC7 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Baculoviral IAP repeat-containing protein 7; Baculoviral IAP repeat-containing protein 7 30kDa subunit; KIAP; KIAP LIVIN ML-IAP MLIAP RNF50; Kidney inhibitor of apoptosis protein; Livin; livin inhibitor of apoptosis; Melanoma inhibitor of apoptosis protein; ML-IAP; RING finger protein 50; RING-type E3 ubiquitin transferase BIRC7; Truncated livin
Gene Aliases: BIRC7; KIAP; LIVIN; ML-IAP; MLIAP; RNF50; UNQ5800/PRO19607/PRO21344
UniProt ID: (Human) Q96CA5
Entrez Gene ID: (Human) 79444
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.