Novus Biologicals
Manufacturer Code:NB300264
Catalog # NB300264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from a sequence between residue 4775 and the C-terminus (residue 4829) of human Baculoviral IAP repeat-containing protein 6 using the numbering given in Swiss-Prot entry Q9NR09 (GeneID 57448). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APOLLON; APOLLON baculoviral IAP repeat containing 6 baculoviral IAP repeat-containing 6 baculoviral IAP repeat-containing protein 6 BRUCE FLJ13786 KIAA1289FLJ13726 Ubiquitin-conjugating BIR domain enzyme apollon ubiquitin-conjugating BIR-domain enzyme apollon; baculoviral IAP repeat-containing 6; Baculoviral IAP repeat-containing protein 6; BIR repeat-containing ubiquitin-conjugating enzyme; BRUCE; RING-type E3 ubiquitin transferase BIRC6; Ubiquitin-conjugating BIR domain enzyme apollon; ubiquitin-conjugating BIR-domain enzyme apollon
Gene Aliases: APOLLON; BIRC6; BRUCE; KIAA1289
UniProt ID: (Human) Q9NR09
Entrez Gene ID: (Human) 57448
Molecular Function: enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.