Invitrogen
Manufacturer Code:PA5109448
Catalog # PIPA5109448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-253 of human BIN3 (NP_061158.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700015F07Rik BIN3 Bridging integrator 3; Bridging integrator 3; bridging integrator-3
Gene Aliases: 1700015F07Rik; BIN3
UniProt ID: (Human) Q9NQY0, (Rat) Q68FW8, (Mouse) Q9JI08
Entrez Gene ID: (Human) 55909, (Rat) 361065, (Mouse) 57784
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.