Proteintech
Manufacturer Code:201861AP150UL
Catalog # 50-173-2756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BIN3 Fusion Protein Ag14091 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BIN3 bridging integrator 3; Bridging integrator 3; bridging integrator-3
Gene Aliases: 1700015F07Rik; BIN3
UniProt ID: (Human) Q9NQY0, (Rat) Q68FW8, (Mouse) Q9JI08
Entrez Gene ID: (Human) 55909, (Rat) 361065, (Mouse) 57784
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.