Proteintech
Manufacturer Code:142451AP150UL
Catalog # 50-172-9313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | BIN2 Fusion Protein Ag5502 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BIN2 BRAP 1 BRAP1 bridging integrator 2; breast cancer associated protein BRAP1; Breast cancer-associated protein 1; Bridging integrator 2; bridging integrator-2
Gene Aliases: BIN2; BRAP-1; BRAP1
UniProt ID: (Human) Q9UBW5
Entrez Gene ID: (Human) 51411
Molecular Function:
G-protein modulator
actin family cytoskeletal protein
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.