Invitrogen
Manufacturer Code:PA582120
Catalog # PIPA582120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human BIN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMPH2 AMPHL SH3P9; Amphiphysin II; Amphiphysin-like protein; box dependant MYC interacting protein 1; Box-dependent myc-interacting protein 1; Bridging integrator 1; Myc box-dependent-interacting protein 1
Gene Aliases: AMPH2; AMPHL; BIN1; SH3P9
UniProt ID: (Human) O00499
Entrez Gene ID: (Human) 274
Molecular Function:
G-protein modulator
actin family cytoskeletal protein
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.