Invitrogen
Manufacturer Code:PA518285
Catalog # PIPA518285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (FNAYYARRLEK) corresponding to the C-terminus amino acids of BCL2L11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; Bcl-2-like protein 11; bcl-2-related ovarian death agonist; Bcl2-interacting mediator of cell death; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator); BIM BCL2L11 BAM BOD BCL2-like 11 (apoptosis facilitator) bcl-2 interacting protein Bim bcl-2-related ovarian death agonist bcl-2 interacting mediator of cell death BIM-alpha6 BIM-beta6 BIM-beta7 BimEL BimL OTTHUMP00000197013
Gene Aliases: BAM; BCL2L11; BIM; BOD
UniProt ID: (Human) O43521
Entrez Gene ID: (Human) 10018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.