Novus Biologicals
Manufacturer Code:NBP222454
Catalog # NBP222454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 23F7 |
Immunogen | Residues 5-195 from human recombinant protein expressed in bacteria |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: apoptic death agonist; apoptic death agonist BH3 interacting domain death agonist BH3-interacting domain death agonist BID isoform ES(1b) BID isoform L(2) BID isoform Si6 desmocollin type 4 FP497 Human BID coding sequence MGC15319 MGC42355 p22 BID; BH3-interacting domain death agonist; BH3-interacting domain death agonist p11; BH3-interacting domain death agonist p13; BH3-interacting domain death agonist p15; BID; BID isoform ES(1b); BID isoform L(2); BID isoform Si6; desmocollin type 4; Human BID coding sequence; p11 BID; p13 BID; p15 BID; p22 BID
Gene Aliases: BID; FP497
UniProt ID: (Human) P55957
Entrez Gene ID: (Human) 637
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.