Invitrogen
Manufacturer Code:PA520559
Catalog # PIPA520559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 13 amino acid peptide from near the carboxy terminus of human BICD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bic-D 2; bicaudal D homolog 2; bicaudal D protein; coiled-coil protein BICD2; cytoskeleton-like bicaudal D protein homolog 2; homolog of Drosophila bicaudal D; Protein bicaudal D homolog 2; RP11-476B13.3 bA526D8.1 bic-D 2 coiled-coil protein BICD2 cytoskeleton-like bicaudal D protein homolog 2 homolog of Drosophila bicaudal D protein bicaudal D homolog 2
Gene Aliases: bA526D8.1; BICD2; KIAA0699; SMALED2
UniProt ID: (Human) Q8TD16, (Mouse) Q921C5
Entrez Gene ID: (Human) 23299, (Mouse) 76895, (Rat) 306809
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.