Novus Biologicals
Manufacturer Code:NBP178734
Catalog # NBP178734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 300 and 350 of human Bicaudal D Homolog 1 using the numbering given in entry NP_001705.2 (GeneID 636). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bic-D 1; Bicaudal D (Drosophila) homolog 1 bicaudal D homolog 1 (Drosophila) BICD Bic-D 1 cytoskeleton-like bicaudal D protein homolog 1 protein bicaudal D homolog 1; bicaudal D homolog 1; cytoskeleton-like bicaudal D protein homolog 1; Protein bicaudal D homolog 1
Gene Aliases: BICD; BICD1
UniProt ID: (Human) Q96G01
Entrez Gene ID: (Human) 636
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.