Abnova
Manufacturer Code:H00000636A01
Catalog # 89-104-150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 70 a.a) partial recombinant protein with GST tag.,BICD1 (NP_001705 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant BICD1.
For Research Use Only
Protein Aliases: Bic-D 1; bicaudal D homolog 1; BICD; cytoskeleton-like bicaudal D protein homolog 1; Protein bicaudal D homolog 1
Gene Aliases: BICD; BICD1
UniProt ID: (Human) Q96G01
Entrez Gene ID: (Human) 636
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.