Abnova
Manufacturer Code:H00023743M02A
Catalog # 89-106-644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E8 |
Immunogen | BHMT2 (NP_0600841 a.a. &bsim 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant BHMT2.
For Research Use Only
Protein Aliases: Betaine--homocysteine S-methyltransferase 2; betaine-homocysteine methyltransferase 2; FLJ20001; S-methylmethionine--homocysteine S-methyltransferase BHMT2; SMM-hcy methyltransferase
Gene Aliases: BHMT2
UniProt ID: (Human) Q9H2M3
Entrez Gene ID: (Human) 23743
Molecular Function: decarboxylase lyase methyltransferase oxidoreductase reductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.