Novus Biologicals
Manufacturer Code:NBP215585
Catalog # NBP215585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human BHMT2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Betaine--homocysteine S-methyltransferase 2; betaine-homocysteine methyltransferase 2; betaine-homocysteine methyltransferase 2 betaine--homocysteine S-methyltransferase 2 EC 2.1.1.5 FLJ20001; S-methylmethionine--homocysteine S-methyltransferase BHMT2; SMM-hcy methyltransferase
Gene Aliases: BHMT2
UniProt ID: (Human) Q9H2M3
Entrez Gene ID: (Human) 23743
Molecular Function: decarboxylase lyase methyltransferase oxidoreductase reductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.