Invitrogen
Manufacturer Code:PA521466
Catalog # PIPA521466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 199 and 406 of BHMT (Uniprot IDNo.Q93088) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Betaine--homocysteine S-methyltransferase 1; BHMT1 betaine--homocysteine S-methyltransferase 1; epididymis secretory sperm binding protein Li 61p
Gene Aliases: BHMT; BHMT1; HEL-S-61p
UniProt ID: (Human) Q93088
Entrez Gene ID: (Human) 635
Molecular Function: decarboxylase lyase methyltransferase oxidoreductase reductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.