Invitrogen
Manufacturer Code:PA5114904
Catalog # PIPA5114904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BHMT corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Betaine--homocysteine S-methyltransferase 1; betaine-homocysteine methyltransferase; betaine-homocysteine methyltransferase betaine--homocysteine S-methyltransferase betaine-homocysteine S-methyltransferase betaine--homocysteine S-methyltransferase 1 BHMT BHMT1 epididymis secretory sperm binding protein Li 61p HEL-S-61p; epididymis secretory sperm binding protein Li 61p
Gene Aliases: BHMT; BHMT1; HEL-S-61p
UniProt ID: (Human) Q93088, (Rat) O09171, (Mouse) O35490
Entrez Gene ID: (Human) 635, (Rat) 81508, (Mouse) 12116
Molecular Function:
decarboxylase
lyase
methyltransferase
oxidoreductase
reductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.