OriGene
Manufacturer Code:TA806394
Catalog # 50-168-1095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5B1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-297 of human BHLHE41 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix domain containing, class B, 3; basic helix-loop-helix family, member e41; bHLHb3; BHLHB3 DEC2 SHARP1 hDEC2; bHLHe41; Class B basic helix-loop-helix protein 3; Class E basic helix-loop-helix protein 41; Differentially expressed in chondrocytes protein 2; Enhancer-of-split and hairy-related protein 1; hDEC2; SHARP-1
Gene Aliases: BHLHB3; BHLHE41; DEC2; hDEC2; SHARP1
UniProt ID: (Human) Q9C0J9
Entrez Gene ID: (Human) 79365
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.